Plasmid isolation, single-strand phage DNA, M-13 isolation kits, E.Z.N.A.® and E-Z 96®
Supplier: OMEGA BIO-TEK
D6900-00 D6900-01
OMEGD6900-00EA 0 SGD
OMEGD6900-00 OMEGD6900-01
Plasmid isolation, single-strand phage DNA, M-13 isolation kits, E.Z.N.A.® and E-Z 96®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
E.Z.N.A.® M13 DNA kits are designed to purify up to 10 μg of single-stranded DNA from up to 3 ml of phage supernatant. Yields of single-stranded DNA obtained using E.Z.N.A.® M13 DNA kits are around 3 to 10 μg and reproducible when the isolations are performed from the same culture. Kits are also available in E-Z 96® DNA plate format.