Digital camera, C-P20CC
Supplier: OPTIKA
C-P20CC
OPTKC-P20CCEA 0 SGD
OPTKC-P20CC
Digital camera, C-P20CC
Cameras
CMOS cooled colour camera for fluorescence applications with high sensitivity and low noise.
  • Peltier-cooled to 45 °C below camera body temperature
  • C-mount connection for microscopes
  • High-speed live view camera for smooth results at maximum resolution
  • Extremely reliable colour fidelity sensor
  • High resolution and long time exposure for vivid details
Recommended for routinary and research level on different observation methods.
Ordering information: Includes a comprehensive software suite.
Delivery information: Equipped with a calibration slideMore Product Information