Dissolved oxygen probe, standard, Orion™ 081010MD
Supplier: Thermo Orion
081010MD
ORIO081010MDEA 0 SGD
ORIO081010MD
Dissolved oxygen probe, standard, Orion™ 081010MD
Electrodes Dissolved Oxygen Electrodes
This versatile standard DO probe for the lab or field allows reliable results no matter what the environment.
  • For use with Star Series meters
  • Response time: 90% of final value in 10 sec.
  • Drift: <1%/day
Order Now
SPECIFICATIONS
Environmentally Preferable