Orion™ RDO™ Dissolved Oxygen Probes
Supplier: Thermo Orion
087010MD
ORIO087010MDEA 0 SGD
ORIO087010MD
Orion™ RDO™ Dissolved Oxygen Probes
Electrodes Dissolved Oxygen Electrodes
These RDO (Rugged Dissolved Oxygen) probes use optical DO technology for problem-free measurement.
  • Measure dissolved oxygen levels without the difficulties of membrane replacement
  • Once-a-year maintenance with a simple cap replacement
  • Different cable lengths available
  • IP 68-rated, waterproof
Can be used with Orion Star meters with RDO input.
Delivery information: Supplied with optical cap, stainless steel sensor guard and calibration sleeve.