Resistor kit for conductivity meters, Orion™
Supplier: Thermo Orion
1010001
ORIO1010001EA 0 SGD
ORIO1010001
Resistor kit for conductivity meters, Orion™
Electrochemistry Meters Conductivity Meters
This resistor calibration kit can be used to verify the performance of Thermo Scientific™ Orion™ Versa Star conductivity meters, Thermo Scientific™ Orion™ Star A series conductivity meters and legacy Orion™ Star series conductivity meters.
  • Conductivity calibration resistor kit with 8 pin MiniDIN connection
Delivery information: Kit includes six resistors that simulate nominal conductance values between 1 µS and 100 mS.