Accessories
Supplier: Thermo Orion
1010003
ORIO1010003EA 0 SGD
ORIO1010003
Accessories
Electrochemistry Meters pH and Multi-Parameter Meters
Spare or replacement universal power adapter for the Thermo Scientific™ Orion™ Star A series (Star A320, Star A220, Star A210, Star A120 and Star A110 series) benchtop and portable meters and legacy Orion™ Star and Star Plus series meters.
110/222 V, 50/60 Hz