pH electrode, Orion™ 8103BNUWP ROSS Ultra™
Supplier: Thermo Orion
8103BNUWP
ORIO8103BNUWPEA 0 SGD
ORIO8103BNUWP
pH electrode, Orion™ 8103BNUWP ROSS Ultra™
Electrodes pH Electrodes
This general purpose, semi-micro tipped combination electrode with top-performance glass body can be used for precise pH determinations in micro volume samples. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day)
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable