pH electrode, Orion™ 8104BNUWP ROSS Ultra™
Supplier: Thermo Orion
8104BNUWP
ORIO8104BNUWPEA 0 SGD
ORIO8104BNUWP
pH electrode, Orion™ 8104BNUWP ROSS Ultra™
Electrodes pH Electrodes
This general purpose combination electrode with toughened bulb and top-performance glass body can be used for precise pH determinations. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is
More Product Information