pH combination electrodes, Orion™ ROSS Ultra™ low maintenance pH/ATC Triode™
Supplier: Thermo Orion
8107BNUMD
ORIO8107BNUMDEA 0 SGD
ORIO8107BNUMD
pH combination electrodes, Orion™ ROSS Ultra™ low maintenance pH/ATC Triode™
Electrodes pH Electrodes
These low maintenance ROSS pH electrodes offer unmatched benefits that users will not find in any other pH electrode. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample
More Product Information