Combination pH electrodes with flat surface tip, Orion™ 8135 ROSS™
Supplier: Thermo Orion
8135BN
ORIO8135BNEA 0 SGD
ORIO8135BN
Combination pH electrodes with flat surface tip, Orion™ 8135 ROSS™
Electrodes pH Electrodes
The ROSS combination flat surface pH electrodes measure pH of soft, moist surfaces — both solid and semi-solid — such as agar gel plates, meats, bread dough and similar samples. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable