Combination pH electrode, Orion™ 815600 ROSS™
Supplier: Thermo Orion
815600
ORIO815600EA 0 SGD
ORIO815600
Combination pH electrode, Orion™ 815600 ROSS™
Electrodes pH Electrodes
ROSS pH electrodes offer unmatched benefits that users will not find in any other pH electrode. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample contamination: ROSS
More Product Information