pH electrode, Orion™ 8156BNUWP ROSS Ultra™
Supplier: Thermo Orion
8156BNUWP
ORIO8156BNUWPEA 0 SGD
ORIO8156BNUWP
pH electrode, Orion™ 8156BNUWP ROSS Ultra™
Electrodes pH Electrodes
This top performance pH electrode can be used to precisely perform pH determinations. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample contamination: ROSS electrodes
More Product Information