pH electrode, Orion™ 8175BNWP ROSS™ Sure-Flow™
Supplier: Thermo Orion
8175BNWP
ORIO8175BNWPEA 0 SGD
ORIO8175BNWP
pH electrode, Orion™ 8175BNWP ROSS™ Sure-Flow™
Electrodes pH Electrodes
This general purpose pH electrode can be used to measure general pH samples as well as viscous or colloidal samples. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • Sure-Flow™: Clog-resistant, easy-to-clean sleeve junction
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,0
More Product Information