Combination pH micro electrode, Orion™ PerpHecT™ ROSS™
Supplier: Thermo Orion
8220BNWP
ORIO8220BNWPEA 0 SGD
ORIO8220BNWP
Combination pH micro electrode, Orion™ PerpHecT™ ROSS™
Electrodes pH Electrodes
This pH electrode is ideal for measuring micro volume samples. A PTFE protective sleeve over the glass stem provides greater durability than conventional micro pH electrodes and allows pH and temperature measurement of samples in 384 well plates or samples.
Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other
More Product Information