Orion™ ROSS Ultra™ Triode™ pH/ATC Combination Electrode
Supplier: Thermo Orion
8302BNUMD
ORIO8302BNUMDEA 0 SGD
ORIO8302BNUMD
Orion™ ROSS Ultra™ Triode™ pH/ATC Combination Electrode
Electrodes pH Electrodes
This Triode™ electrode has a built-in temperature sensor allowing users to measure pH and temperature with only one electrode. Ideal for complex samples, such as biological media, foods and pharmaceuticals. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS
More Product Information