ISE filling solutions, Orion™
Supplier: Thermo Orion
900017 900067 900010 900012 900046 900062 900063 900065 951202 951209
ORIO900017EA 0 SGD
ORIO900017 ORIO900067 ORIO900010 ORIO900012 ORIO900046 ORIO900062 ORIO900063 ORIO900065 ORIO951202 ORIO951209
ISE filling solutions, Orion™
Electrolytes Electrode Filling Solutions
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
  • Enhance the performance and extend the life of ion selective electrodes
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective combination electrodes