Reference electrode, Orion™ 900100 Sure-Flow™
Supplier: Thermo Orion
900100
ORIO900100EA 0 SGD
ORIO900100
Reference electrode, Orion™ 900100 Sure-Flow™
Electrodes ISE Electrodes
This reference half-cell electrode helps to precisely measure with ion-selective electrodes. Ideal for ISE determinations and routine pH measurements with Orion 910100 electrodes.
  • Ag/AgCl reference
  • SureFlow sleeve junction
Reference electrode is recommended for all pH and ion-selective measurements except for those:
- In dilute solutions where the ion to be measured is contained in the filling solution of the Orion 900100
- In solutions where an ion in the filling solution would introduce an electrode interference
- In solutions where a species isMore Product Information