Filling solutions for pH electrodes, Orion™
Supplier: Thermo Orion
900003 900019 510011 810007 900001 900002 900004 900011 910008-WA
ORIO900003EA 0 SGD
ORIO900003 ORIO900019 ORIO510011 ORIO810007 ORIO900001 ORIO900002 ORIO900004 ORIO900011 ORIO910008-WA
Filling solutions for pH electrodes, Orion™
Electrolytes Electrode Filling Solutions
Filling solutions for pH and reference electrodes.
  • ROSS filling solution for use only with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode or double junction reference electrodes are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide
Order Now
SPECIFICATIONS
Environmentally Preferable