Combination pH electrodes, Orion™ Basic
Supplier: Thermo Orion
9146BN
ORIO9146BNEA 0 SGD
ORIO9146BN
Combination pH electrodes, Orion™ Basic
Electrodes pH Electrodes
These pH electrodes are ideal for performing basic pH measurements quickly and reliably. Designed for use with Orion™ Russell meters.
  • Can be used with any meter with BNC input
  • For general purpose applications
Storage requirements: store in Orion pH electrode storage solution 910001.
Six months from date of purchase by buyer or 12 months from date of shipment from seller.