Combination pH electrode, Orion™ 9156BNWP
Supplier: Thermo Orion
9156BNWP
ORIO9156BNWPEA 0 SGD
ORIO9156BNWP
Combination pH electrode, Orion™ 9156BNWP
Electrodes pH Electrodes
This general purpose electrode is ideal for performing routine pH measurements under rugged conditions.
  • Unbreakable body extends beyond the pH sensing glass bulb for protection
  • With waterproof connector
Delivery information: Supplied with 60 ml bottle of Orion electrode fill solution.
Order Now
SPECIFICATIONS
Environmentally Preferable