3-in-1 pH/ATC probes, Orion™ Triode™
Supplier: Thermo Orion
9157BNMD
ORIO9157BNMDEA 0 SGD
ORIO9157BNMD
3-in-1 pH/ATC probes, Orion™ Triode™
Electrodes pH Electrodes
These versatile 3-in-1 pH/ATC probes can be used in either the field or a plant.
  • Type reference: Ag/AgCl
  • Ag/AgCl internal reference system, sealed reference
  • Refillable
Delivery information: Supplied with one 60 ml bottle of filling solution.
Order Now
SPECIFICATIONS
Environmentally Preferable