pH electrode, low resistance, Orion™ 9162BNWP
Supplier: Thermo Orion
9162BNWP
ORIO9162BNWPEA 0 SGD
ORIO9162BNWP
pH electrode, low resistance, Orion™ 9162BNWP
Electrodes pH Electrodes
This low resistance electrode is ideal for use in viscous solutions.
  • Silver/silver chloride (Ag/AgCl) reference: High accuracy and broad temperature range
  • Rugged bulb, easy to clean
  • Waterproof BNC connector
  • Cap diameter: 16 mm
Warranty: 6 months from date of purchase by Buyer or 12 months from date of shipment from Seller, whichever is earlier.
Order Now
SPECIFICATIONS
Environmentally Preferable