pH electrode, Orion™ Sure-Flow 91-72
Supplier: Thermo Orion
9172BNWP
ORIO9172BNWPEA 0 SGD
ORIO9172BNWP
pH electrode, Orion™ Sure-Flow 91-72
Electrodes pH Electrodes
Chemical resistant glass body combination electrode for clog-free measurements of soil, sludge, colloids, and viscous material.
  • Sure-Flow junction: Free flowing liquid-to-liquid junction provides the most stable, drift-free measurements
  • Refillable Ag/AgCl combination pH electrode
  • Waterproof BNC connector
  • Can be used on any pH meter with a BNC connection