Ion selective electrodes, nitrate, Orion™
Supplier: Thermo Orion
9307BNWP 9707BNWP
ORIO9307BNWPEA 0 SGD
ORIO9307BNWP ORIO9707BNWP
Ion selective electrodes, nitrate, Orion™
Electrodes ISE Electrodes
These nitrate electrodes are designed for measuring nitrate in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • Combination electrode 9707BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable