Ion selective electrode, surfactant, Orion™
Supplier: Thermo Orion
9342BN
ORIO9342BNEA 0 SGD
ORIO9342BN
Ion selective electrode, surfactant, Orion™
Electrodes ISE Electrodes
This surfactant half cell is a potentiometric sensor for end point detection in titrations of anionic or cationic surfactants in a solution. A potentiometric sensor, it is a surfactant-detecting electrode in anionic or cationic titration which is specifically designed for use on the Orion 960 Autochemistry system.
Can be used on any ISE or mV meter with a BNC connection. Must be used with the double junction reference electrode.