Ion selective electrodes, cyanide, Orion™
Supplier: Thermo Orion
9406BN 9606BNWP
ORIO9406BNEA 0 SGD
ORIO9406BN ORIO9606BNWP
Ion selective electrodes, cyanide, Orion™
Electrodes ISE Electrodes
These electrodes are designed for measuring cyanide ions in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode does not require a separate reference
More Product Information