Ion selective electrodes, cupric, Orion™
Supplier: Thermo Orion
9429BN 9429SC 9629BNWP
ORIO9429BNEA 0 SGD
ORIO9429BN ORIO9429SC ORIO9629BNWP
Ion selective electrodes, cupric, Orion™
Electrodes ISE Electrodes
These electrodes are designed for measuring cupric ions in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode ORIO9629BNWP does not require a separate
More Product Information