Orion™ 9502BNWP Carbon Dioxide Electrode
Supplier: Thermo Orion
9502BNWP
ORIO9502BNWPEA 0 SGD
ORIO9502BNWP
Orion™ 9502BNWP Carbon Dioxide Electrode
Electrodes ISE Electrodes
This electrode measures carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately.
  • Gas-sensing combination electrode
  • With waterproof BNC connector