Ion selective electrode, High Performance, ammonia, Orion™
Supplier: Thermo Orion
9512HPBNWP
ORIO9512HPBNWPEA 0 SGD
ORIO9512HPBNWP
Ion selective electrode, High Performance, ammonia, Orion™
Electrodes ISE Electrodes
The high performance ammonia ion selective electrode (ISE) allows fast, simple, economical, and accurate measurements of dissolved ammonia in aqueous solutions.
  • Response time of 1 minute in sample concentrations of 1 ppm and higher; fast results make it easy to combine functionality with autosamplers
  • Keying feature on the electrode body prevents membrane from twisting or breaking during assembly
  • Choice of preassembled outer body with membrane cap or loose membranes
  • New wastewater applications developed using known addition and direct measurement techniques
More Product Information