Ion selective electrodes, cadmium, Orion™
Supplier: Thermo Orion
9648BNWP
ORIO9648BNWPEA 0 SGD
ORIO9648BNWP
Ion selective electrodes, cadmium, Orion™
Electrodes ISE Electrodes
These electrodes are designed for measuring cadmium ions in aqueous solutions quickly, simply and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode (9648BNWP) does not require a separate reference
More Product Information