Ion selective electrodes, chlorine, Orion™
Supplier: Thermo Orion
9770BNWP
ORIO9770BNWPEA 0 SGD
ORIO9770BNWP
Ion selective electrodes, chlorine, Orion™
Electrodes ISE Electrodes
These electrodes are designed for measuring total residual chlorine quickly and accurately.
  • All forms of total residual chlorine are measured
  • Combination electrode does not require a separate reference electrode, making it convenient to use
Ordering information: The electrode with screw cap is supplied without cable.