pH electrodes, micro, Orion
Supplier: Thermo Orion
9810BN
ORIO9810BNEA 0 SGD
ORIO9810BN
pH electrodes, micro, Orion
Electrodes pH Electrodes
pH electrodes with BNC connector and 1 m cable.
  • pH range: 0 to 14
  • Material: Glass