pH electrode, micro, Orion™ 9826BN
Supplier: Thermo Orion
9826BN
ORIO9826BNEA 0 SGD
ORIO9826BN
pH electrode, micro, Orion™ 9826BN
Electrodes pH Electrodes
This long length combination electrode can be used to accurately measure pH in NMR cells.
  • Made of very thin, delicate glass
  • Refillable
Order Now
SPECIFICATIONS
Environmentally Preferable