pH electrode, micro, Orion™ 9863BN
Supplier: Thermo Orion
9863BN
ORIO9863BNEA 0 SGD
ORIO9863BN
pH electrode, micro, Orion™ 9863BN
Electrodes pH Electrodes
This bevel needle-tipped 16-gauge glass body combination electrode can be used to measure pH by piercing septa.
  • Made of very thin, delicate glass
  • Single junction Ag/AgCl
  • Refillable