Printer, Orion™ Star A series
Supplier: Thermo Orion
STARA-106
ORIOSTARA-106EA 2570 SGD
ORIOSTARA-106
Printer, Orion™ Star A series
Printers Document Printers
This compact ink ribbon printer with built-in paper cutter can be used with Orion Star A210, Star A220 and Star A320 series meters; Dual Star meters; Versa Star and Versa Star Pro meters.
  • USB or Ethernet on-board interface
  • Serial, parallel, USB, Ethernet, Bluetooth and WiFi interfaces options
  • Cutter rated for 1 800 000 cuts
SDK Available for iOS and Android operating systems.
Certifications: FCC, RoHS, Energy Star certified.
Delivery information: Supplied with one ink ribbon, one paper roll, meter interface cable and power cords (110 V US/Japan, 2More Product Information