pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Supplier: Thermo Orion
STARA2110 STARA2115
ORIOSTARA2110EA 0 SGD
ORIOSTARA2110 ORIOSTARA2115
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Electrochemistry Meters pH and Multi-Parameter Meters
Easy to use and budget friendly instrument for a wide range of applications and advanced pH analysis in the lab. Ideal for simple, routine pH, mV and temperature measurements.
  • Performs up to a 5-point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Eliminate the need for magnetic stirring bars using the meter-controlled stirrer probe to uniformly mix solutions
  • Data log collects and
More Product Information