pH/mV/ORP/°C meters, bench top, Orion Star™ A211

Supplier: Thermo Orion
STARA2110 STARA2115
ORIOSTARA2110EA 0 SGD
ORIOSTARA2110 ORIOSTARA2115
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Electrochemistry Meters pH and Multi-Parameter Meters
Easy to use and budget friendly instrument for a wide range of applications and advanced pH analysis in the lab. Ideal for simple, routine pH, mV and temperature measurements.

  • Performs up to a 5-point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Eliminate the need for magnetic stirring bars using the meter-controlled stirrer probe to uniformly mix solutions
  • Data log collects and
More Product Information
Order Now

SPECIFICATIONS

Model A211
Environmentally Preferable
pH range −2,000...+20,000
pH resolution 0,1 / 0,01 / 0,001
pH accuracy ±0,002
pH calibration Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
mV range ±2000,0
mV resolution 0,1
mV accuracy ±0,2 mV or ±0,05% of reading whichever is greater
Temperature range −5,0...+105,0 °C
Temperature resolution 0,1 °C
Temperature accuracy ±0,1 °C
Temperature compensation Automatic or manual
Display Graphic LCD with backlight
Battery type 4×AA (sold separately)
Battery life 800 hours
Power supply AC adapter or 4 AA batteries (800 hours battery life)
Compliance IP 54
Operating temperature range 5...45 °C
Data logging - log function Manual / Automatic with AUTO-READ™
Data logging capacity 2000
Memory Non volatile memory preserves data log, calibration log and meter settings
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Connections BNC, reference pin, 8-pin mini-DIN
Outputs RS232, USB
Input BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
Calibration pH with calibration editing option, relative mV, ORP and temperature
Calibration points 1 - 5
W×D×H 180×240×110 mm
Weight 0,9 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR