Syringe filters, Acrodisc®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
4602
PALL4602EA 527 SGD
PALL4602
Syringe filters, Acrodisc®, Supor® membrane
Filters Syringe Filters
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
  • Superior flow rates and higher throughputs than competitive devices
  • Low protein binding to minimize sample loss
  • Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
  • A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
  • Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
  • Reduces mycoplasma with the
More Product Information