Membrane disc filters
Supplier: Cytiva (Formerly Pall Lab)
66149 66155
PALL66149EA 0 SGD
PALL66149 PALL66155
Membrane disc filters
Filters Membrane Filters
Strong, chemically resistant membranes for air monitoring and sampling in aggressive environments.
  • Ensures gravimetric determinations with low tare mass
  • Ideal for filtration of gas and/or organic solvents
  • Supported membranes offer increased durability for hostile testing environments or acid-aerosol monitoring
Teflo membrane: PTFE with PMP support ring. Typical aerosol retention: 1 and 2 µm: 99,99%; 3 µm: 99,79%.
TF (PTFE) membrane: PTFE on a PP support.
Typical aerosol retention: 0,5; 1 and 2 µm: 99,99%