Centrifugal devices, Nanosep® and Nanosep® MF
Supplier: Cytiva (Formerly Pall Lab)
OD010C34
PALLOD010C34EA 1020 SGD
PALLOD010C34
Centrifugal devices, Nanosep® and Nanosep® MF
Filters Centrifugal Filters
These devices are used for simple, reliable concentrating and desalting of 50 to 500 μl samples. Select the low protein binding Omega™ membrane for ultrafiltration applications. The Bio-Inert® membrane is ideal for microfiltration of biological solutions. The GHP membrane is ideal for sample preparation in analytical chemistry (HPLC, GC, IC and MS). It is suitable for aqueous, organic and aggressive organic samples, and for rapid processing of samples.
  • Rapid processing of samples
  • Typical recoveries are greater than 90%
Housing: Low binding PP.