Cell culture medium, Pantum 386
Supplier: PAN BIOTECH
P04-00386
PANBP04-00386EA 0 SGD
PANBP04-00386
Cell culture medium, Pantum 386
Cell Culture Media
Pantum are ready-to-use growth media which contain purified plasma proteins and lipids, such as serum albumin and cholesterol, specific growth factors, components of soybean extract, an iron transport protein and enriched trace elements.
  • The new formulations result in stable cell growth under defined culture conditions
  • No addition of serum or growth factors is necessary
  • Stored at −20 °C
Pantum 386 is a modified formulation of DMEM and specially developed to optimize the growth of epithelial cell lines.