Cell culture medium, Pantum 586A
Supplier: PAN BIOTECH
P04-00586
PANBP04-00586EA 0 SGD
PANBP04-00586
Cell culture medium, Pantum 586A
Cell Culture Media
Pantum medium are ready-to-use growth media which contain purified plasma proteins and lipids, such as serum albumin and cholesterol, specific growth factors, components of soybean extract, an iron transport protein and enriched trace elements.
  • The new formulations result in stable cell growth under defined culture conditions
  • No addition of serum or growth factors is necessary
Pantum 586A is a modification of Iscove`s MEM and particularly suited for culture of adherent cells to stimulate their growth.