Serum-free media, Panserin 604
Supplier: PAN BIOTECH
P04-710604
PANBP04-710604EA 0 SGD
PANBP04-710604
Serum-free media, Panserin 604
Cell Culture Media
Panserin 604 is a complete medium ready for use for the cultivation of transfected and non-transfected CHO-cells (Chinese Hamster Ovary) in an adherent culture.
  • Highly enriched medium for the fast growth and culture of adherent CHO-cells
Based on Glasgow medium, trace elements, albumin, cholesterol, soya-lipids, vitamins hormones and attachment factors were added to the medium.
Suitable for the cultivation of transfected and non-transfected CHO-cells in adherent culture (e.g. roller cultures).