Water, Cell Culture Grade, sterile-filtered

Supplier: PAN BIOTECH

Synonyms: Hydrogen oxide

P04-99200B
PANBP04-99200BEA 0 SGD
PANBP04-99200B
Water, Cell Culture Grade, sterile-filtered
Water

Maximum efficiency without loss of quality reagents are tested according to the highest possible quality standards. All liquid reagents are dissolved according to in-house specifications, sterilised and filtered at 0,2 μm. Before final release, the reagents undergo extensive quality tests example sterility, pH value, osmosis.


  • Store from 2 °C to room temperature

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR