Penicillin/Streptomycin/Amphotericin B mixture 10000 U/ml: 10 mg/ml: 25 µg/ml in saline (0.85%)

Supplier: PAN BIOTECH
Danger

P06-07325
PANBP06-07325EA 0 SGD
PANBP06-07325
Penicillin/Streptomycin/Amphotericin B mixture 10000 U/ml: 10 mg/ml: 25 µg/ml in saline (0.85%)
Penicillin/Streptomycin/Amphotericin B

  • Penicillin:10000 U/ml
  • Streptomycin: 10 mg/ml
  • Amphotericin B: 25 µg/ml


Even though cells should be cultured without the addition of antibiotics or antifungal drugs, cell culture media are not only a perfect breeding ground for your cell culture but also for many contaminants, such as bacteria and fungi. In certain cases the addition of antibiotics is necessary (for unique cell lines or to remove already existing contamination).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR