MEM vitamin supplement 100X
Supplier: PAN BIOTECH
P08-41001P
PANBP08-41001PEA 0 SGD
PANBP08-41001P
MEM vitamin supplement 100X
Cell Culture Supplements and Additives
The availability of vitamins is extremely important for the cells, since the cells usually cannot synthesise the vitamins.
  • Stored at 2 to 8 °C
Vitamins act as co-factors for many enzymes and are essential for their function. The absence of vitamins in culture may lead to decrease in cell growth, cell death or loss of productivity.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more