CELLkit
Supplier: PAN BIOTECH
P09-1004K
PANBP09-1004KEA 0 SGD
PANBP09-1004K
CELLkit
Cells
Kit contains Endopan 300 SL and Human Umblical Vein Endothelial Cells (HUVEC/TERT66).
  • Sterile
Endopan 300 SL is serum free complete medium for human endothelial cells and optimised for the maintenance and expansion of endothelial cells. Endopan 300 SL kit is provided with serum substitute and supplements in separate sterile packing. This will enable the user to prepare a medium for special applications.
HUVEC are valuable in vitro model that has contributed to major insights in regulation of endothelial cell function and angiogenesis or the pathophysiology of anthrosclerosis and plaque formation.
They can be used to study vascularisation in response to hypoxic conditions in tumor or ischemic tissues, co-culture with telomerised MSCs (ASC/TERT1) as enhanced in vitro model for studying vascular biology, assessment of interaction with leukocytes and macrophages,study of inflammation.