VWR®, Transport bag with biohazard symbol with fast-zip closure and pocket for documents
Supplier: VWR Collection
PBIB29738.CEA 147 SGD
PBIB29738.C
VWR®, Transport bag with biohazard symbol with fast-zip closure and pocket for documents
Bags Sample Bags
Bags made from PE, transparent.
  • For the transport of samples
  • Pressure seals
  • Complete with separate external pocket for documentation
  • Closure with pressure band