VWR® SAS Super IAQ Air Samplers
Supplier: VWR Collection
PBIB90578EA 0 SGD
PBIB90578 PBIB90715
VWR® SAS Super IAQ Air Samplers
Samplers Air Samplers
The SAS Super IAQ provides the same efficiency as recorded by sampling tests conducted by Centre for Applied Microbiology & Research (CAMR-UK).
  • Microbiological air monitoring solution for agri-food and environmental operators
  • Includes all accessories required for microbiological air sampling
  • Easy to use and time saving, low operating cost solution
Designed specifically for the Indoor Air Quality (IAQ) sector, this SAS Super instrument is lightweight and durable and able to operate in any environment, therefore, suitable for environmentalMore Product Information