VWR® Safe-Tamper System
Supplier: VWR Collection
PBIB91985.BEA 0 SGD
PBIB91985.B
VWR® Safe-Tamper System
Bags Sample Bags
Anti-tamper disposable bags, in PE, with double pocket for internal hospital biological sample transportation. Double pocket bags: Two separate compartments for lab document and specimen. It is possible to customise the bags.
  • Transparent bags, yellow coloured to identify leakage of fluid and for a better visibility of the contents
  • Adhesive closure to prevent tampering and to preserve patient privacy
  • Printed biohazard logo and instructions
  • The bag is destroyed by opening it, preventing reuse and avoiding any risk of cross-contamination